Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Iron(III) chloride, anhydrous ≥97%

Supplier: Pfaltz & Bauer
Danger
Synonyms: Ferric chlorideIron trichloride

F00710-500G
101187-974EA 289 USD InStock
101187-974
Iron(III) chloride, anhydrous ≥97%
Iron (III) chloride
Synonyms: Ferric perchloride anhydrous


Formula: FeCl₃
MW: 162.21 g/mol
Boiling Pt: 316 °C
Melting Pt: 300…306 °C
Density: 2.804
MDL Number: MFCD00011005
CAS Number: 7705-08-0
UN: 1773
ADR: 8,III
Order Now
Specification Test Results
Appearance Brown crystals
Solubility Soluble in Water, Alcohol, Ether, Acetone
pH Value 2 for 0.1 M aqueous solution

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty