Would you like to log in again?
Go Direct™ Gas Pressure
Supplier: VERNIER SOFTWARE & TECH SE
GDX-GP
470166-508EA 98.99 USD InStock
470166-508
Go Direct™ Gas Pressure
Data Loggers
The Go Direct Gas Pressure Sensor measures the absolute pressure of a gas.
  • Investigate Boyle’s law and Charles’ law
  • Investigate grip strength and muscle fatigue (using Gas Pressure Bulb, not included)
  • Measure the production of oxygen gas produced when hydrogen peroxide is destroyed by the enzyme catalase
  • Monitor plant transpiration
Use Go Direct Gas Pressure Sensor to monitor gas pressure in a variety of experiments. It records accurate absolute pressure readings relative to the perfect vacuum reference point inside the sensor, allowing you to go below atmospheric pressure or up to 400 kPa. Easily change the displayed units to any one of seven options (kPa, mmHg, inHg, mbar, psi, atm, torr). Includes a syringe, tubing, and stoppers to ease setup for experiments such as Boyle’s law.
The Go Direct robust rechargeable battery offers long battery life and provides always-ready operation. Monitor battery life directly from our free Graphical Analysis™ 4 app. Should the battery run low, simply connect the sensor to the charging cable and keep on collecting—no interruptions or inconvenience to you or your students.
Our Go Direct™ Gas Pressure Sensor connects directly to your mobile device, Chromebook, or computer using our free Graphical Analysis™ 4 app—no additional equipment or software purchases are necessary. Go Direct sensors can be used wired via USB or wirelessly via Bluetooth, allowing you to choose the solution best for your classroom or laboratory.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty