Would you like to log in again?
Go Direct™ Radiation Monitor
Supplier: VERNIER SOFTWARE & TECH SE
GDX-RAD
470166-534EA 218.99 USD InStock
470166-534
Go Direct™ Radiation Monitor
Data Loggers
The Go Direct Radiation Monitor detects alpha, beta, gamma, and X-ray radiation.
  • The Go Direct Radiation Monitor can be used in a variety of experiments
  • Determine the half-life of an isotope
  • Investigate radiation shielding
  • Detect background radiation sources
Use the Go Direct Radiation Monitor to explore radiation statistics, measure the rate of nuclear decay, and monitor radon progeny. This easy-to-use sensor consists of a Geiger-Mueller tube mounted in a small, rugged, plastic case. A thin window protected by a metal screen allows alpha radiation to be detected, along with beta and gamma. Each new detection is indicated by a flashing red LED indicator and can also be accompanied by an optional audio signal.
The Go Direct robust rechargeable battery offers long battery life and provides always-ready operation. Monitor battery life directly from our free Graphical Analysis™ 4 app. Should the battery run low, simply connect the sensor to the charging cable and keep on collecting—no interruptions or inconvenience to you or your students.
Our Go Direct™ Radiation Monitor connects directly to your mobile device, Chromebook, or computer using our free Graphical Analysis™ 4 app—no additional equipment or software purchases are necessary. Go Direct sensors can be used wired via USB or wirelessly via Bluetooth, allowing you to choose the solution best for your classroom or laboratory.
Delivery information: Includes Go Direct radiation monitor, micro USB cable.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty