Would you like to log in again?
Go Direct™ Force and Acceleration
Supplier: VERNIER SOFTWARE & TECH SE
GDX-FOR
470166-538EA 118.99 USD InStock
470166-538
Go Direct™ Force and Acceleration
Data Loggers
A three-axis accelerometer with a stable and accurate force sensor that measures forces as small as ±0.1 N and up to ±50 N.
  • Go Direct force and acceleration can be used in a variety of experiments
  • Investigate Newton’s Third Law by connecting the hooks of two force sensors with a rubber band
  • Use the force sensor to pull an object across a surface to measure frictional forces
  • Attach the force sensor to the centripetal force apparatus to measure centripetal force and acceleration simultaneously
  • Place sensors on Dynamics carts to investigate forces and accelerations in collisions
The Go Direct force and acceleration sensor includes a force sensor, three-axis accelerometer, and three-axis gyroscope. Take it on a roller coaster, swing, or slide. Suspend several Go Direct force and acceleration sensors from the ceiling to perform a 3-D vector force experiment, or attach a string to the hook and whirl it in a horizontal or vertical circle. In wireless mode, your imagination is the only limiting factor!
The Go Direct robust rechargeable battery offers long battery life and provides always-ready operation. Monitor battery life directly from our free Graphical Analysis™ 4 app. Should the battery run low, simply connect the sensor to the charging cable and keep on collecting—no interruptions or inconvenience to you or your students.
Our Go Direct™ Force and Acceleration Sensor connects directly to your mobile device, Chromebook, or computer using our free Graphical Analysis™ 4 app—no additional equipment or software purchases are necessary. Go Direct sensors can be used wired via USB or wirelessly via Bluetooth, allowing you to choose the solution best for your classroom or laboratory.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty