Would you like to log in again?
Vernier Spectrophotometer Optical Fiber
Supplier: VERNIER SOFTWARE & TECH SE
VSP-FIBER
470225-926EA 78.99 USD InStock
470225-926
Vernier Spectrophotometer Optical Fiber
Cables
Easy to use fiber optic cable for use with Vernier Go Direct SpectroVis.
  • Works with:
  • Go Direct™ SpectroVis Plus Spectrophotometer
  • Vernier Fluorescence/UV-VIS Spectrophotometer
  • Vernier UV-VIS Spectrophotometer
This is an optical fiber accessory designed exclusively for emission spectrum experiments with Vernier-branded spectrometers. Simply insert the unit into the cuvette holder and point the tip of the 1 m optical fiber at a light source. The unit is keyed for safety. It has a wavelength range from 350 nm to 900 nm.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty