Would you like to log in again?
SP Wilmad-LabGlass Round Bottom Flasks with Threaded Standard Taper Joint, SP Industries
Supplier: SP Industries
ML-1110-608
82003-040EA 68.96 USD InStock
82003-040
SP Wilmad-LabGlass Round Bottom Flasks with Threaded Standard Taper Joint, SP Industries
Flasks Round Bottom Flasks
These single neck round bottom flasks have a threaded Standard Taper 14/10 outer joint.
  • Borosilicate glass
Delivery information: Supplied with one 20-400 open top nylon screw cap and one size 014 Viton® O-ring.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty