Would you like to log in again?
Ward's® Essentials Green Glucose Production Kit
Supplier: Avantor
Ward's®
470227-320KT 74.59 USD InStock
470227-320
Ward's® Essentials Green Glucose Production Kit
Educational Classroom Kits and Activities
Measure photosynthesis and respiration using algae beads.
  • Hands on lab activity
  • Kit aligned to Next Generation Science Standards MS-LS1-6 and MS-ETS1-1
  • Instructions and materials included for 15 groups
  • Middle school
A simple organism, algae beads, are used to determine the rates of photosynthesis and respiration in a colorimetric assay. Using a pH indicator, test the effects of light on these important cellular processes.
Ordering information: Coupon is included for redemption of perishable materials. Redeem by mail, phone, fax, email or online.
Caution: WARNING: CHOKING HAZARD–Product contains small parts, small balls, or marbles. Not for children under 3 yrs.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty