Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Ward's® Essentials Torque Feeler Kit
Supplier: EISCO SCIENTIFIC LLC
Ward's®
VWRGP00002
470225-368KT 43.99 USD InStock
470225-368
Ward's® Essentials Torque Feeler Kit
Educational Classroom Kits and Activities
Feel how the torque changes when the position of the mass changes!
  • Hands on lab activity
  • Kit aligned to Next Generation Science Standards MS-PS2-2
  • Instructions and materials included for 1 group
  • Middle school
With this kit the user can experience significantly increasing torque by moving a mass set down a rod while trying to balance the main rod. It is a tricky and fun demonstration that will leave students with a lasting conceptual understanding.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty