Would you like to log in again?
Pharma-65 Tubing, Silicone, DuPont
Supplier: DuPont
4008344 4008345 4077978 4090602 4077179 4008346
89198-888PK 437.9 USD InStock
89198-888 89198-886 89198-880 89198-890 89198-884 89198-882
Pharma-65 Tubing, Silicone, DuPont
Tubing Transfer Tubing
Pharmaceutical grade silicone tubing for transferring of ultra-pure liquids, air or steam in pharmaceutical and biotechnological manufacturing processes.
  • Excellent flexibility and low extractables
  • Easily sterilized
  • High resiliency
  • Complete traceability
  • No impaired taste or odor
Certifications: Meets or exceeds 21 CFR 177.2600, GMPs, USP Class VI and 3A Sanitary Standard procedures and European Pharmacopoeia Monograph 3.1.9. silicone elastomer for closures and tubing.
Packaging: Standard package includes 15-meter coil, double bagged polyethylene bags in a cardboard box.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty