Would you like to log in again?
Screw Thread SureStop (AVCS) Vials, Thermo Scientific
Supplier: Thermo Scientific
C5000-1 C5000-1W
82428-550PK 45.64 USD InStock
82428-550 82428-548
Screw Thread SureStop (AVCS) Vials, Thermo Scientific
Vials
These screw thread vials are built with Advanced Vial Closure System (AVCS).
  • Fill vials easily with the wide neck opening
  • Requires 6mm diameter micro-inserts
  • Nearly eliminate septa push-through and improve sealing
Vials are made from superior quality 33 expansion borosilicate clear (Type 1, Class A) glass, and meet all US, EU, and JPN Pharmacopia standards.
This innovation not only prevents septa push through, but provides a stop point for the cap to prevent over- or under-tightening and provide an optimal, consistent seal every time.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty