Would you like to log in again?
Surgifoam® Absorbable Gelatin Powder Kit, ETHICON
Supplier: Johnson & Johnson
ETH1979
10611-622CS 4378.7 USD InStock
10611-622
Surgifoam® Absorbable Gelatin Powder Kit, ETHICON
First Aid Supplies Surgical Drapes
Absorbable gelatin powder is a sterile porcine intended for hemostatic use by applying to a bleeding surface.
  • Store dry at controlled room temperature of 15 to 30 °C (59 to 86 °F)
This gelatin powder kit controls bleeding by controlling the amount of thrombin or saline added. Conforms to irregular surfaces to stop bleeding fast.
Ordering information: Kit includes syringe and tip.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty