Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(-)-Dibenzoyl-L-tartaric acid monohydrate ≥98.0%

Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms: (2R,3R)-(-)-Di-O-benzoyl-tartaric acid monohydrate

D1584-100GM
10860-484EA 242 USD InStock
10860-484
(-)-Dibenzoyl-L-tartaric acid monohydrate ≥98.0%
(-)-Dibenzoyl-L-tartaric acid monohydrate
GHS hazard : Exclamation mark


Formula: C₁₈H₁₄O₈·1H₂O
MW: 376.33 (358.31 anhy)
Storage Temperature: Ambient
MDL Number: MFCD00149119
CAS Number: 62708-56-9
Order Now
Specification Test Results
Purity (Neutralization Titration) 98.0 % min.
Specific Rotation @ 20°C -106.0 to -111.0
Water 4.7 - 5.1%

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty