Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(-)-Epigallocatechin gallate

Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms: (-)-cis-3,4-Dihydro-5,7-dihydroxy-2-(3,4,5-trihydroxyphenyl)-1(2H)-benzopyran-3-yl Gallate(2R,3R)-5,7-Dihydroxy-2-(3,4,5-trihydroxyphenyl)chroman-3-yl 3,4,5-trihydroxybenzoateEGCG

E1154-50MG
700008-480EA 113.22 USD InStock
700008-480
(-)-Epigallocatechin gallate
(-)-Epigallocatechin gallate
(-)-Epigallocatechin Gallate is an antioxidant polyphenol flavonoid isolated from green tea. Its possible benefit as a nutritional chemopreventive agent for cancer, atherosclerosis, and neurodegenerative diseases is generating increased scientific interest. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available


Formula: C₂₂H₁₈O₁₁
MW: 458.38 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00075940
CAS Number: 989-51-5
Order Now
Specification Test Results
Appearance Cream Colored Po
Purity 95% min.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty