Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(+)-Dibenzoyl-D-tartaric acid monohydrate ≥95.0%

Supplier: SPECTRUM CHEMICAL MFG CORP

D1582-100GM
10860-506EA 224 USD InStock
10860-506
(+)-Dibenzoyl-D-tartaric acid monohydrate ≥95.0%
(+)-Dibenzoyl-D-tartaric acid monohydrate
GHS hazard : Exclamation mark


Formula: C₁₈H₁₄O₈·1H₂O
Storage Temperature: Ambient
MDL Number: MFCD00150722
CAS Number: 80822-15-7
Order Now
Specification Test Results
Purity (HPLC; Area %) 95.0 % min.
Water 4.3 - 5.1 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty