Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Clomipramine hydrochloride ≥98.0% USP

Supplier: SPECTRUM CHEMICAL MFG CORP

C1376-1KGBL C1376-100GM C1376-5GM C1376-25GM
75810-798EA 576 USD InStock
75810-798 75810-796 75810-794 75810-792
Clomipramine hydrochloride ≥98.0% USP
Clomipramine hydrochloride
Clomipramine Hydrochloride, USP has a trade name of Anafranil and has several uses in medicine including the treatment of panic disorder, major depressive disorder, and chronic pain. The USP grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

GHS hazard : Exclamation mark


Formula: C₁₉H₂₄Cl₂N₂
Storage Temperature: Ambient
MDL Number: MFCD00069234
CAS Number: 17321-77-6
Order Now
Specification Test Results
Assay (Dried Basis) 98.0 - 102.0%
pH 3.5 - 5.0
Loss On Drying max. 1.0%
Organic Impurities, Procedures 1 and 2 To Pass Test
Residue On Ignition max. 0.1%

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty