Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Copper(II) sulfate pentahydrate, BiotechGrade

Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms: Cupric sulfate pentahydrateCopper sulfate pentahydrateCopper sulphate pentahydrateCupric sulphate pentahydrate

C1421-12KG C1421-45KGBL C1421-500GM C1421-2.5KG
75810-814EA 203 USD InStock
75810-814 75810-812 75810-810 75810-808
Copper(II) sulfate pentahydrate, BiotechGrade
Copper (II) sulphate pentahydrate
Cupric Sulfate, Pentahydrate, BiotechGrade also known as Copper(II) Sulfate is a chemical compound that exists as a salt or crystal and differs in the degree of hydration. It uses very from use as a herbicide to use as a dye. It is also used in Benedict's solution, Fehling's solution and Biuret reagent. Spectrum offers highly pure reagents suitable for biochemical research and analysis. The critical parameters involved are absence of inhibitors such as traces of heavy metals as well as biochemical function tests for enzymes, coenzymes and enzyme substrates.

GHS hazard : Skull and crossbones;Exclamation mark
UN3077


Formula: CuSO₄·5H₂O
MW: 249.69 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00149681
CAS Number: 7758-99-8
UN: 3077
ADR: 9,III
Order Now
Specification Test Results
Appearance Blue, Crystallin
Odor Odorless
Assay (CuSO₄) 98.5 - 100.5 %
Loss On Drying @ 250°C 33.0 - 36.5%
Solubility (5G in 50ML) To Pass Test

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty