Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-(-)-Pantolactone

Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms: (R)-(-)-Dihydro-3-hydroxy-4,4-dimethyl-2(3H)-furanoneD-(-)-2-Hydroxy-3,3-dimethyl-gamma-butyrolactone(R)-3-Hydroxy-4,4-dimethyldihydrofuran-2(3H)-oneD-(-)-Pantoyl Lactone

P2154-25GM P2154-100GM P2154-5GM
700008-420EA 77 USD InStock
700008-420 700008-418 700008-416
D-(-)-Pantolactone
D-(-)-Pantolactone
D-(-)-Pantoyl Lactone is used in facial cleansing products containing exfoliating particles (excluding products for acne).

Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.


Formula: C₆H₁₀O₃
MW: 130.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00005392
CAS Number: 599-04-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty