Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Disodium hydrogen phosphate heptahydrate ≥98.0%, crystals USP

Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms: Sodium phosphate dibasic heptahydrateSodium monohydrogen phosphate heptahydrateDisodium hydrogenorthophosphate 7-hydrateDisodium hydrogenorthophosphate heptahydrateSodium phosphate dibasic 7-hydrateDisodium phosphate (heptahydrate)di-Sodium hydrogen phosphate 7-hydrateSodium monohydrogen phosphate 7-hydrateSodium hydrogen phosphate heptahydrate

SO140-12KG SO140-50KGBL SO140-500GM SO140-2.5KG
95035-872EA 249 USD InStock
95035-872 75812-432 95035-870 89050-384
Disodium hydrogen phosphate heptahydrate ≥98.0%, crystals USP
di-Sodium hydrogen phosphate heptahydrate
Dibasic Sodium Phosphate, Heptahydrate, Crystal, USP is used as a saline laxative to treat constipation or to clean the bowel before a colonoscopy. Spectrum Chemical USP products, including all USP Sodium Phosphate products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities


Formula: HNa₂O₄P·7H₂O
MW: 268.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00149180
CAS Number: 7782-85-6
Order Now
Specification Test Results
Assay (Dried Basis) 98.0 - 100.5 %
Loss On Drying 43.0 - 50.0 %
Insoluble Matter max. 0.4 %
Chloride (Cl) max. 0.06 %
Sulfate (SO₄) max. 0.2 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty