Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(+)-cis-Diltiazem hydrochloride ≥98.0% USP

Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms: (2S,3S)-5-(2-(Dimethylamino)ethyl)-2-(4-methoxyphenyl)-4-oxo-2,3,4,5-tetrahydrobenzo[b][1,4]thiazepin-3-yl acetate hydrochloride(+)-cis-3-Acetoxy-5-(2-dimethylaminoethyl)-2,3-dihydro-2-(4-methoxyphenyl)-1,5-benzothiazepin-4(5H)-one Hydrochloride(2S,3S)-(+)-3-Acetoxy-2,3-dihydro-5-[2-(dimethylamino)ethyl]-2-(4-methoxyphenyl)-1,5-benzothiazepin-4(5H)-one HydrochlorideDiltiazem Hydrochloride

D1163-5GM D1163-100GM D1163-25GM
95032-936EA 220 USD InStock
95032-936 95032-934 95032-930
(+)-cis-Diltiazem hydrochloride ≥98.0% USP
(+)-cis-Diltiazem hydrochloride
Diltiazem Hydrochloride, USP is a calcium channel blocker used to prevent chest pain (angina). All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

GHS hazard : Skull and crossbones;Exclamation mark;Health hazard


Formula: C₂₂H₂₆N₂O₄S·HCl
MW: 450.98 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00069252
CAS Number: 33286-22-5
Order Now
Specification Test Results
Assay (Dried Basis) 98.0 - 102.0%
Specific Rotation [A]D +110° to +116°
Loss On Drying max. 0.5%
Residue On Ignition max. 0.1%
Heavy Metals (As Pb) max. 20 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty