Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethyl violet

Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms: Basic Violet 4

ET120-25GM
700010-182EA 82 USD InStock
700010-182
Ethyl violet
Ethyl violet
Ethyl Violet is used as a tissue stain and as a reagent for the extractive-spectrophotometric determination of anionic surfactants.

Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.


Formula: C₃₁H₄₂ClN₃
MW: 560.29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011765
CAS Number: 2390-59-2
Order Now
Specification Test Results
Visual Transition Intervals pH 0.0 (Yellow)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty