Would you like to log in again?
Gellan gum
Supplier: SPECTRUM CHEMICAL MFG CORP
G1139-125GM G1139-25KGBL G1139-500GM G1139-2.5KG
700011-584EA 7049 USD InStock
700011-584 700011-586 700011-580 700011-582
Gellan gum
Gellan gum
Gellan Gum is a polysaccharide produced by a bacterium named Pseudomonas elodea and is used primarily as a gelling agent, alternative to agar, in microbiological culture.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty