Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Guaifenesin ≥98.0% USP

Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms: 3-(2-Methoxyphenoxy)-1,2-propanediol3-(2-Methoxyphenoxy)propane-1,2-diolGuaiacol glyceryl ether

GU102-50KGBL GU102-100GM GU102-2.5KG GU102-25GM GU102-12KG GU102-500GM GU102-04
95033-460EA 103 USD InStock
95033-460 700003-604 95033-462 CA11024-342 75811-288 700003-602 95033-458
Guaifenesin ≥98.0% USP
Guaifenesin
Guaifenesin, USP is an expectorant. It helps loosen congestion in your chest and throat. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

GHS hazard : Exclamation mark


Formula: C₁₀H₁₄O₄
MW: 198.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00016873
CAS Number: 93-14-1
Order Now
Specification Test Results
Assay (Dried Basis) 98.0 - 102.0 %
Organic Impurities To Pass Test
Loss On Drying max. 0.5 %
Heavy Metals max. 25 ppm
Identification To Pass Test

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty