Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Jojoba oil

Supplier: SPECTRUM CHEMICAL MFG CORP

J1052-500ML J1052-200LTB J1052-100ML J1052-25ML
700000-180EA 384 USD InStock
700000-180 700000-174 700000-176 700000-178
Jojoba oil
Jojoba oil
Jojoba Oil is known as a fungicide and has been used for mildew control. It can also be used as a whale oil cetyl alcohol replacement.

Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.


Density: 0.870
Storage Temperature: Refrigerator
MDL Number: MFCD00131426
CAS Number: 61789-91-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty