Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Melanin (synthetic)

Supplier: SPECTRUM CHEMICAL MFG CORP

M1674-250MG M1674-1GM
700011-972EA 446 USD InStock
700011-972 700011-970
Melanin (synthetic)
Melanin
Melanin, Synthetic is used in personal care sun protection products like as a sunscreens.

Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.


Formula: C₁₈H₁₀N₂O₄
MW: 318.288 g/mol
Storage Temperature: Low Temperature Freezer (–40°C)
MDL Number: MFCD00131581
CAS Number: 8049-97-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty