Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Menthyl acetate ≥96%

Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms: rel-(1R,2S,5R)-2-Isopropyl-5-methylcyclohexyl acetate

M1678-100ML M1678-25ML
700004-552EA 220 USD InStock
700004-552 700004-554
Menthyl acetate ≥96%
Menthyl acetate
Menthyl Acetate is a natural monoterpene which contributes to the smell and flavor of peppermint.

Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.


Formula: C₁₂H₂₂O₂
MW: 198.3019 g/mol
Density: 0.922
Storage Temperature: Ambient
MDL Number: MFCD00062975
CAS Number: 89-48-5
Order Now
Specification Test Results
Assay 96 - 100%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty