Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L-Methionine ≥98.5% USP

Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms: (S)-(-)-2-Amino-4-(methylthio)butyric acidL(-)-MethionineMethionine(S)-(-)-2-Amino-4-(methylthio)butanoic acidH-Met-OH(S)-(-)-Methionine(S)-2-Amino-4-(methylthio)butanoic acid

M1441-500GM M1441-100GM M1441-25KGBL M1441-25GM M1441-1KG
95034-036EA 470 USD InStock
95034-036 95034-032 75811-660 95034-030 75811-662
L-Methionine ≥98.5% USP
L-Methionine
Methionine, USP is used to prevent liver damage in acetaminophen poisoning. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


Formula: C₅H₁₁NO₂S
MW: 149.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00063097
CAS Number: 63-68-3
Order Now
Specification Test Results
Assay (Dried Basis) 98.5 - 101.5 %
Specific Rotation +22.4 to +24.7°
pH (1 in 100) 5.6 - 6.1
Loss On Drying max. 0.3 %
Related Compounds To Pass Test

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty