Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methocarbamol ≥98.5% USP

Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms: Carbamic Acid 2-Hydroxy-3-(2-methoxyphenoxy)propyl Ester2-Hydroxy-3-(2-methoxyphenoxy)propyl Carbamate

M1088-25GM M1088-25KGBL M1088-2.5KG M1088-1KG M1088-500GM M1088-100GM
75811-558EA 289 USD InStock
75811-558 75811-554 75811-556 75811-560 75811-552 95034-438
Methocarbamol ≥98.5% USP
Methocarbamol
Methocarbamol, USP is used to treat muscle spasms and pain. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

GHS hazard : Exclamation mark


Formula: C₁₁H₁₅NO₅
MW: 241.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00057662
CAS Number: 532-03-6
Order Now
Specification Test Results
Assay (Dried Basis) 98.5 - 101.5 %
Loss On Drying max.0.5 %
Residue On Ignition max. 0.1 %
Heavy Metals max. 0.002 %
Organic Impurities To Pass Test

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty