Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Acetyl-L-histidine monohydrate

Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms: Ac-His-OH·H2OAc-His-OH.H2O

A3258-25GM A3258-5GM A3258-1GM
700011-614EA 109 USD InStock
700011-614 700011-610 700011-612
N-Acetyl-L-histidine monohydrate
N-Acetyl-L-histidine monohydrate
Nalpha-Acetyl-L-histidine, Monohydrate , a histidine derivative, is a prominent biomolecule in brain, retina and lens of poikilothermic vertebrates.

Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.


Formula: C₈H₁₁N₃O₃·H₂O
MW: 215.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00149320
CAS Number: 39145-52-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty