Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid ≥98.5% USP

Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms: Naproxen(S)-2-(6-Methoxynaphthalen-2-yl)propanoic acid(S)-(+)-6-Methoxy-alpha-methyl-2-naphthaleneacetic Acid

N1034-12KGBL N1034-500GM N1034-1KG N1034-100GM N1034-25KGBL
75811-744EA 15904 USD InStock
75811-744 75811-742 95034-708 95034-706 95034-712
(S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid ≥98.5% USP
(S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid
Naproxen, USP is a non-steroidal anti-inflammatory drug. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

GHS hazard : Skull and crossbones;Exclamation mark;Health hazard
UN2811


Formula: C₁₄H₁₄O₃
MW: 230.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00010500
CAS Number: 22204-53-1
UN: 2811
ADR: 6.1,III
Order Now
Specification Test Results
Assay (Dried Basis) 98.5 - 101.5 %
Specific Rotation +83.0 to +89.5
Chromatographic Purity To Pass Test
Loss On Drying max. 0.5 %
Heavy Metals max. 0.002 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty