Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Solvent black 5

Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms: 2-Amino-N,N-dipropylacetamide hydrochlorideNigrosin soluble in spiritNigrosin B, alcohol soluble

NI110-25GM NI110-100GM
700010-316EA 72 USD InStock
700010-316 700010-314
Solvent black 5
Solvent black 5


Formula: C₈H₁₉ClN₂O
Storage Temperature: Ambient
MDL Number: MFCD00071316
CAS Number: 11099-03-9
Order Now
Specification Test Results
Absorption Max. (In Ethanol) Approx. 565 NM

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty