Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phenylarsine oxide

Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms: OxophenylarsineArzenePAO

P1580-5GM P1580-1GM P1580-25GM
10841-846EA 260.1 USD InStock
10841-846 10841-844 10841-842
Phenylarsine oxide
Phenylarsine oxide
GHS hazard : Skull and crossbones
UN3465


Formula: C₆H₅AsO
MW: 168.02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00001990
CAS Number: 637-03-6
UN: 3465
ADR: 6.1,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty