Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Prilocaine hydrochloride ≥98.0% USP

Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms: 2-(Propylamino)-N-(o-tolyl)propionamide Hydrochloride

PR102-5GM PR102-1KGBL PR102-25GM PR102-100GM PR102-500GMB PR102-500GMB
95035-344EA 1991 USD InStock
95035-344 95035-336 95035-346 95035-340 95035-342 700007-478
Prilocaine hydrochloride ≥98.0% USP
Prilocaine hydrochloride
Prilocaine Hydrochloride, USP is a local anesthetic used most often for infiltration anesthesia in dentistry. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

GHS hazard : Exclamation mark


Formula: C₁₃H₂₀N₂O
MW: 256.77 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00079279
CAS Number: 1786-81-8
Order Now
Specification Test Results
Assay (Dried Basis) 98.0 - 102.0 %
Loss On Drying max.0.3 %
Residue On Ignition max. 0.1 %
Heavy Metals max. 0.002 %
Organic Impurities To Pass Test

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty