Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pullulan

Supplier: SPECTRUM CHEMICAL MFG CORP

P1830-100GM P1830-500GM P1830-50KGBL P1830-25GM
700013-212EA 141 USD InStock
700013-212 700013-214 700013-216 700013-210
Pullulan
Pullulan
Pullulan, a polysaccharide polymer, is used in the manufacture of edible films used in oral hygiene products.

Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.


Formula: (C₆H₁₂O₅)n
Storage Temperature: Ambient
MDL Number: MFCD00081940
CAS Number: 9057-02-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty