Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Quinidine sulfate Dihydrate ≥99.0% USP

Supplier: SPECTRUM CHEMICAL MFG CORP

QU105-50GM QU105-10GM QU105-100GM
95035-520EA 545 USD InStock
95035-520 95035-522 95035-524
Quinidine sulfate Dihydrate ≥99.0% USP
Quinidine sulfate Dihydrate
Quinidine Sulfate, Dihydrate, USP is an antimalarial and antiarrhythmic drug used to treat or prevent many types of irregular heartbeats. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

GHS hazard : Exclamation mark


Formula: C₄₀H₄₈N₄O₄.H₂O₄S·2H₂O
Storage Temperature: Ambient
MDL Number: MFCD00149346
CAS Number: 6591-63-5
Order Now
Specification Test Results
Assay (Anhydrous Basis) 99.0 - 101.0%
Specific Rotation [A]D +275° to +288°
Water Determination 4.0 - 5.5%
Residue After Ignition max. 0.1%
Organic Impurities To Pass Test

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty