Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Salicylic acid ≥98.0%, powder USP

Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms: o-Hydroxybenzoic acid2-Hydroxybenzoic acid

SA130-500GM SA130-2.5KG SA130-12KG SA130-25KGBL
95035-610EA 806 USD InStock
95035-610 700006-388 75812-416 95035-612
Salicylic acid ≥98.0%, powder USP
Salicylic acid
Salicylic Acid, Powder, USP is a topical application used to treat skin conditions that involve scaling or overgrowth of skin cells such as psoriasis, dandruff, corns, calluses, and warts.

All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


Formula: (HO)C₆H₄CO₂H
MW: 138.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00002439
CAS Number: 69-72-7
https://us-legacy.vwr.com/stibo/web/std.lang.all/99/00/749900.jpg
Order Now
Specification Test Results
Assay (Dried Basis) 98.0 - 102.0 %
Loss On Drying max.0.5 %
Residue On Ignition max. 0.05 %
Chloride (Cl) max. 0.014 %
Sulfates (SO₄) max. 0.02 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty