Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium acetate, anhydrous ≥99.0%, BiotechGrade

Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms: Acetic acid sodium salt

S1104-12KG S1104-500GM-CS S1104-2.5KG S1104-500GM
75811-898EA 840 USD InStock
75811-898 75812-764 75812-124 75812-122
Sodium acetate, anhydrous ≥99.0%, BiotechGrade
Sodium acetate
Sodium Acetate, Anhydrous, BiotechGrade is a colorless salt that has a wide range of uses. It is used as part of buffer solution to help keep a near constant pH. Spectrum offers highly pure reagents suitable for biochemical research and analysis. The critical parameters involved are absence of inhibitors such as traces of heavy metals as well as biochemical function tests for enzymes, coenzymes and enzyme substrates.


Formula: H₃CCOONa
MW: 82.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00012459
CAS Number: 127-09-3
Order Now
Specification Test Results
Assay 99.0 %
pH of a 5% Solution @ 25°C 7.0 - 9.2
Solubility (10 G in 100 ML) Practically Clea
Loss On Drying @ 120°C max. 1.0 %
Insoluble Matter max. 0.005 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty