Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium acetate, anhydrous ≥99.0% USP

Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms: Acetic acid sodium salt

SO104-500GM SO104-500GM SO104-50KGBL SO104-12KG SO104-2.5KG SO104-2.5KG
700004-296EA 777 USD InStock
CA700004-296 700004-296 700004-294 CA700004-300 700004-300 75812-418
Sodium acetate, anhydrous ≥99.0% USP
Sodium acetate
Sodium Acetate, Anhydrous, USP is used as an electrolyte replenisher in isosmotic solution for parenteral replacement of acute losses of extracellular fluid without disturbing normal electrolyte balance. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


Formula: H₃CCOONa
MW: 82.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00012459
CAS Number: 127-09-3
Order Now
Specification Test Results
Assay (Dried Basis) 99.0 - 101.0 %
pH of a 3% Solution @ 25°C 7.5 - 9.2
Loss On Drying max. 1.0 %
Insoluble Matter max. 0.05 %
Chloride (Cl) max. 350 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty