Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium fluoride ≥98.5%, powder USP, Ph. Eur., BP

Supplier: SPECTRUM CHEMICAL MFG CORP

SO167-500GM SO167-12KG SO167-45KGBL SO167-2.5KG SO167-125GM
95031-320EA 1122 USD InStock
95031-320 75812-438 95031-318 95031-322 75812-440
Sodium fluoride ≥98.5%, powder USP, Ph. Eur., BP
Sodium fluoride
Sodium Fluoride, Powder, USP, EP, BP is used to prevent cavities making teeth stronger and more resistant to decay caused by acid and bacteria. Toothpaste being the preferred delivery method. All Spectrum Chemical USP, EP, BP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

GHS hazard : Skull and crossbones;Exclamation mark;Health hazard
UN1690


Formula: NaF
MW: 41.99 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003524
CAS Number: 7681-49-4
UN: 1690
ADR: 6.1,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/06/30/16640630.jpg
Order Now
Specification Test Results
Assay (NaF; Dried Basis) 98.5 - 100.5%
Acidity or Alkalinity To Pass Test
Appearance of Solution To Pass Test
Loss On Drying max. 0.5%
Fluosilicate To Pass Test

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty