Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Strontium acetate hemihydrate

Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms: Strontium Acetate Hemihydrate

S2417-100GM S2417-500GM S2417-2.5KG S2417-45KGBL
700004-362EA 82 USD InStock
700004-362 700004-366 700004-364 700004-368
Strontium acetate hemihydrate
Strontium acetate hemihydrate
Strontium Acetate is a compound used in some toothpastes to desensitize teeth to extremes of hot and cold, sweets, acids, or contact.

Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.


Formula: C₈H₁₄O₉Sr₂
MW: 429,43 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00198064
CAS Number: 543-94-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty