Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Talc FCC

Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms: SoapstoneMagnesium silicate monohydrateTalcum venetum

T1002-12KG T1002-45KGBL
75812-514EA 627.41 USD InStock
75812-514 75812-516
Talc FCC
Talc
Talc, FCC is approved as a carrier for food colorings and as a separating agent. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

GHS hazard : Health hazard


Formula: Mg₃O₁₁Si₄·1H₂O
MW: 379.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00792903
CAS Number: 14807-96-6
Order Now
Specification Test Results
Acid-Soluble Substances max. 2.5 %
Arsenic (As) max.3 mg/kg
Free Alkali max. 1 %
Lead (Pb) max. 5 mg/kg
Loss On Drying max. 0.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty