Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Triolein ≥99%

Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms: Glycerol TrioleateGlycerine trioleate

T3108-5GM
700011-948EA 276 USD InStock
700011-948
Triolein ≥99%
Triolein
Triolein, a symmetrical triglyceride, is also known as glyceryl trioleate. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.


Formula: C₅₇H₁₀₄O₆
MW: 885.45 g/mol
Density: 0,91
Storage Temperature: Freezer
MDL Number: MFCD00137563
CAS Number: 122-32-7
Order Now
Specification Test Results
Assay (GC) 99 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty