Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Water USP, purified

Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms: Dihydrogen oxide

W1014-200LTBL W1014-20LT W1014-4LT W1014-20LT
700002-172EA 628 USD InStock
700002-172 CA700002-172 75812-604 95036-568
Water USP, purified
Water
Purified Water, USP has gone through a purification process to attain a level of purity. 

All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


Formula: H₂O
MW: 18.02 g/mol
Density: 1.000
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty