Would you like to log in again?
Dounce Tissue Homogenizer, Biovision
Supplier: BioVision
AB286839-1UNITS
10013-840EA 599.71 USD InStock
10013-840
Dounce Tissue Homogenizer, Biovision
Homogenizers Tissue Grinders
The tissue grinder is used for homogenizing delicate tissues such as intestines, liver, heart and cells.
  • Used in conjunction with Cell Fractionation and assays for grinding tissue samples
  • Designed for hand operation
  • Hollow handle permits packing with ice
  • Features expanded reservoir and pouring lip
  • Manually operated, interchangeable pestles
Tubes are made of low extractable borosilicate glass. Clearance between pestle and grinding chamber is 0.09 to 0.16 mm.
Certifications: Glass meets ASTM type 1 Class A and USP Type 1 standards.
Caution: For research use only! Not for use in humans.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty