Would you like to log in again?
Prolystica™ 2X Concentrate Alkaline Detergent, STERIS®
Supplier: Steris
Prolystica™
1C34P3
89169-048EA 1512.55 USD InStock
89169-048
Prolystica™ 2X Concentrate Alkaline Detergent, STERIS®
Cleaning Agents Detergents and Surface Cleansers
Prolystica 2X Concentrate Alkaline Detergent is an alkaline pH liquid detergent used in the removal of a broad range of soils from surgical instruments and medical devices in the manual and automated cleaning process.
  • Effectively removes blood, fat, mucus and other hard-to-remove-soils on surgical instruments and medical devices
  • Broad-spectrum substrate compatibility including stainless steel, plastics and soft metals
  • Safe for use on aluminum container systems
It can be used for stainless steel surgical instruments, select soft metals and apparatus where blood, protein, carbohydrates, mucus and lipids form hard to remove soils.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty