Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Z-Leu-Leu-Glu-7-amino-4-methylcoumarin

Supplier: ADIPOGEN CORP MS
Synonyms: Z-Leu-Leu-Glu-AMC

AG-CP3-0022-M001 AG-CP3-0022-M005
102988-496EA 48.46 USD InStock
102988-496 102988-498
Z-Leu-Leu-Glu-7-amino-4-methylcoumarin
Z-Leu-Leu-Glu-7-amino-4-methylcoumarin
Fluorogenic substrate for the peptidylglutamyl-peptide hydrolysing (PGPH) (caspase-like) activity of the 20S proteasome. Excitation: 380nm. Emission: 460nm.


Formula: C₃₅H₄₄N₄O₉
MW: 664.756 g/mol
MDL Number: MFCD01074989
CAS Number: 348086-66-8
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty