Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

ABD-F [=4-(Aminosulfonyl)-7-fluoro-2,1,3-benzoxadiazole]

Supplier: ADIPOGEN CORP MS
Synonyms: 4-(Aminosulfonyl)-7-fluoro-2,1,3-benzoxadiazole7-Fluoro-2,1,3-Benzoxadiazole-4-Sulfonamide

CDX-F0016-M010
102987-950EA 210.46 USD InStock
102987-950
ABD-F [=4-(Aminosulfonyl)-7-fluoro-2,1,3-benzoxadiazole]
ABD-F [=4-(Aminosulfonyl)-7-fluoro-2,1,3-benzoxadiazole]
Highly reactive fluorogenic reagent for the labeling of thiols for HPLC. ABD-F is nonfluorescent until reacted with thiols and therefore can be used to quantitate thiols in solution, as well as thiols separated by HPLC or TLC. ABD-F also reportedly reacts slowly with the hydroxy group of some tyrosine residues as well as amino groups in some proteins, forming products that are nonfluorescent but can be detected by absorbance at 385 nm.


Formula: C₆H₄FN₃O₃S
MW: 217.17 g/mol
MDL Number: MFCD00036687
CAS Number: 91366-65-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty