Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Fluoro-7-nitrobenzofurazan

Supplier: ADIPOGEN CORP MS
Synonyms: NBD-F4-Fluoro-7-nitro-[2,1,3]-benzoxadiazole

CDX-F0017-M025 CDX-F0017-M250
102987-952EA 211.85 USD InStock
102987-952 102987-954
4-Fluoro-7-nitrobenzofurazan
4-Fluoro-7-nitrobenzofurazan
NBD fluoride is similar to NBD chloride except that the former is much more reactive toward amines. Both reagents are nonfluorescent until they react with primary or secondary amines to produce a fluorescent product. NBD chloride and NBD fluoride have been extensively used as derivatizing reagents for chromatography analysis of amino acids and low molecular weight amines.


Formula: C₆H₂FN₃O₃
MW: 183.1 g/mol
MDL Number: MFCD00010196
CAS Number: 29270-56-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty