Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Alcian blue 8GX 50% (by UV/Vis)

Supplier: ADIPOGEN CORP MS
Synonyms: C.I. 74240C.N. AnAn-6Ingrain Blue 1Alcian blue 8 GX

CDX-A0201-L500 CDX-A0201-DD25
102989-262EA 211.85 USD InStock
102989-262 102989-264
Alcian blue 8GX 50% (by UV/Vis)
Alcian blue 8 GX
Alcian blue 8GX is primarily used for demonstrating acid mucopolysaccharides, which it does quite selectively. In recent years there has been some difficulty obtaining the dye. Alcian Blue 8GX is also suitable for detection of glycoproteins on nitrocellulose and in PAGE gels. There are other dyes similar to alcian blue, although not necessarily belonging to the same class of dyes. Alcian green has gained some popularity, and alcian yellow has been suggested, especially in a method for demonstrating Helicobacter. They are used for acid mucopolysaccharides in the same way as alcian blue. They have no particular advantage for demonstrating mucin. According to the Merck Index the groups marked X split off during staining.


Formula: C₅₆H₆₈Cl₄CuN₁₆S₄
MW: 1298.86 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00010720
CAS Number: 75881-23-1
Order Now
Specification Test Results
Source/Host Synthetic.
Purity 50% (Dye content) (UV/Vis)
Appearance/Color Blue liquid.
Solubility Soluble in 1% in water solution (3% acetic acid).

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty