Would you like to log in again?
Calbiochem® 1,5-Dansyl-Glu-Gly-Arg Chloromethyl Ketone, DiHCl
Supplier: EMD Millipore
Calbiochem®
251700-5MG
80056-266EA 447.49 USD InStock
80056-266
Calbiochem® 1,5-Dansyl-Glu-Gly-Arg Chloromethyl Ketone, DiHCl
Proteins and Peptides
Effective irreversible inhibitor of uPA, Factor Xa, and urokinase
  • Primary target: UPA (100 nM for factor Xa)
  • Protect from moisture
  • Off-white solid
Reconstitute at 1 mg/mL in 10 mM HCl. Dilute with physiological buffers just prior to use.
Fluorophore allows direct monitoring of interactions of labeled enzyme. It does not inhibit tPA.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty