Would you like to log in again?
Microvette® 500 Capillary Blood Collection Tubes, Sarstedt
Supplier: Sarstedt
Microvette®
20.1346.100 20.1344
103218-893PK 108.34 USD InStock
103218-893 103252-123
Microvette® 500 Capillary Blood Collection Tubes, Sarstedt
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
Microvette® tubes with round bottom inner tube are ideal for the collection of blood using the gravity-flow principle
  • Flat Base, Cylindrical Inner Tube 500 µl
  • Optimized tube geometry ensures thorough sample mixing
  • Quadruple-threaded cap can be attached to the tube bottom during collection to prevent mixup or loss
  • Shelf life and batch number on each tube
  • Supplied with printed label
Tubes are made of polypropylene and caps are made of HDPE.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty